Learn More
Invitrogen™ RPL14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593174
Description
Immunogen sequence: MVFRRFVEVG RVAYVSFGPH AGKLVAIVDV IDQNRALVDG PCTQVRRQAM PFKCMQLTDF ILKFPHSAHQ KYVRQAWQKA DINTKWAATR WAKKIEARER KAKMTDFDRF KVMKAKKMRN RIIKNEVKKL QKAALLKASP KKAPGTKGTA AAAAAAAAAK VPAKKITAAS KKAPAQKVPA QKATGQKAAP APKAQKGQKA PAQKAPAPKA SGKKA; Positive Samples: HL-60, HeLa, 293T, Jurkat, HepG2, Mouse ovary.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L14E family of ribosomal proteins. It contains a basic region-leucine zipper (bZIP)-like domain. The protein is located in the cytoplasm. This gene contains a trinucleotide (GCT) repeat tract whose length is highly polymorphic; these triplet repeats result in a stretch of alanine residues in the encoded protein. Transcript variants utilizing alternative polyA signals and alternative 5'-terminal exons exist but all encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Aoki M., Diabetes 45:157-164(1996). Tanaka M., Biochem. Biophys. Res. Commun. 243:531-537(1998). Yoshihama M., Genome Res. 12:379-390(2002).
Specifications
RPL14 | |
Polyclonal | |
Unconjugated | |
RPL14 | |
3100001N19Rik; 60S ribosomal protein L14; AA407502; AL022816; CAG-ISL 7; CAG-ISL-7; CTG-B33; hRL14; L14; Large ribosomal subunit protein eL14; MGC88594; ribosomal protein L14; RL14; RPL14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
65043, 67115, 9045 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50914, Q63507, Q9CR57 | |
RPL14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human RPL14 (NP_0039643). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.