Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549954 Supplier Invitrogen™ Supplier No. PA549954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RPL15 protein.

May play a role as co-chaperone of the Hsp70 family proteins HSPA1A, HSPA1B and HSPA8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rpl15
Gene Accession No. P61313, P61314, Q9CZM2
Gene Alias 2510008H07Rik; 60S ribosomal protein L15; DBA12; EC45; L15; Large ribosomal subunit protein eL15; ribosomal protein L15; RPL10; RPL15; RPLY10; RPYL10; TCBAP0781
Gene Symbols Rpl15
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human RPL15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245981, 6138, 66480
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.