Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583815
Description
Immunogen sequence: RAHGRINPYM SSPCHIEMIL TEKEQIVPKP EEEVAQKKKI SQKKLK.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L22P family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as rpL23 because the encoded protein shares amino acid identity with ribosomal protein L23 from Halobacterium marismortui; however, its official symbol is RPL17. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring downstream C18orf32 gene.
Specifications
RPL17 | |
Polyclonal | |
Unconjugated | |
Rpl17 | |
60S ribosomal protein L17; 60S ribosomal protein L23; Amino acid starvation-induced protein; ASI; gene encoding putative NFkB activating protein; L17; L23; Large ribosomal subunit protein uL22; MGC117162; PD-1; ribosomal protein L17; RPL17; RPL23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6139 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P18621 | |
Rpl17 | |
Recombinant protein corresponding to Human RPL17. Recombinant protein control fragment (Product #RP-99539). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction