Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588025 Supplier Thermo Scientific Supplier No. PA588025
Add to Cart
Add to Cart

RPL18 Polyclonal Antibody

RPL18 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RPL18
Gene Accession No. P35980, Q07020
Gene Alias 60S ribosomal protein L18; L18; Large ribosomal subunit protein eL18; ribosomal protein L18; RPL18; Rpl18a
Gene Symbols RPL18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-188 of human RPL18 (NP_0009701).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19899, 6141
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.