Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL18A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297781PCP
Description
RPL18A Polyclonal antibody specifically detects RPL18A in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
RPL18A | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
60S ribosomal protein L18a, ribosomal protein L18a, ribosomal protein L18a-like protein | |
Produced in rabbits immunized with E. coli-derived Human RPL18A fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
6142 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction