Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL18A Monoclonal Antibody (6G6G10)
GREENER_CHOICE

Catalog No. PIMA517167
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517167 Supplier Invitrogen™ Supplier No. MA517167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17167 targets RPL18A in indirect ELISA, IHC and WB applications and shows reactivity with Human and Mouse samples. The MA5-17167 immunogen is purified recombinant fragment of human RPL18A (amino acids: 50-176) expressed in E. Coli. MA5-17167 detects RPL18A which has a predicted molecular weight of approximately 20.8kDa.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL18A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6G6G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RPL18A
Gene Accession No. P62717, Q02543
Gene Alias 2510019J09Rik; 60S ribosomal protein L18a; L18A; Large ribosomal subunit protein eL20; ribosomal protein L18a; ribosomal protein L18a-like protein; Rpl18a
Gene Symbols RPL18A
Host Species Mouse
Immunogen Purified recombinant fragment of human RPL18A (amino acids: 50-176) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6142, 76808
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.