Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL18A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103541
Description
Antibody detects endogenous levels of total RPL18A.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
Specifications
RPL18A | |
Polyclonal | |
Unconjugated | |
RPL18A | |
2510019J09Rik; 60S ribosomal protein L18a; L18A; Large ribosomal subunit protein eL20; ribosomal protein L18a; ribosomal protein L18a-like protein; Rpl18a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
290641, 6142, 76808 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62717, P62718, Q02543 | |
RPL18A | |
A synthesized peptide derived from human RPL18A(Accession Q02543), corresponding to amino acid residues G65-A115. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction