Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579935
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579935 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579935 Supplier Invitrogen™ Supplier No. PA579935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human A2780 whole cell, human 293T whole cell, human Jurkat whole cell, rat brain tissue, rat liver tissue, mouse brain tissue, mouse liver tissue. ICC/IF: U20S cell. Flow: U20S cell, A431 cell.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L19E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL19
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene RPL19
Gene Accession No. P84098, P84099, P84100
Gene Alias 60S ribosomal protein L19; DKFZp779D216; FLJ27452; HGNC:10312; L19; Large ribosomal subunit protein eL19; MGC71997; ribosomal protein L19; RPL19
Gene Symbols RPL19
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human RPL19 (132-170aa FKNKRILMEHIHKLKADKARKKLLADQAEARRSKTKEAR).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19921, 6143, 81767
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.