Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561479 Supplier Invitrogen™ Supplier No. PA561479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGTRYMFSRP FRKHGVVPLA TYMRIYKKGD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL21
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpl21
Gene Accession No. P46778
Gene Alias 60S ribosomal protein L21; 8430440E03Rik; HYPT12; L21; Large ribosomal subunit protein eL21; ribosomal protein L21; ribosomal protein L21 pseudogene 16; Rpl21; RPL21P16; similar to ribosomal protein L21
Gene Symbols Rpl21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL21. Recombinant protein control fragment (Product #RP-100807).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.