Learn More
Invitrogen™ RPL21 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | RPL21 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Rpl21 |
| Gene Accession No. | P46778 |
| Gene Alias | 60S ribosomal protein L21; 8430440E03Rik; HYPT12; L21; Large ribosomal subunit protein eL21; ribosomal protein L21; ribosomal protein L21 pseudogene 16; Rpl21; RPL21P16; similar to ribosomal protein L21 |
| Gene Symbols | Rpl21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.