Learn More
Invitrogen™ RPL21 Polyclonal Antibody

Description
Immunogen sequence: MTNTKGKRRG TRYMFSRPFR KHGVVPLATY MRIYKKGDIV DIKGMGTVQK GMPHKCYHGK TGRVYNVTQH AVGIVVNKQV KGKILAKRIN VRIEHIKHSK SRDSFLKRVK ENDQKKKEAK EKGTWVQLKR QPAPPREAHF VRTNGKEPEL LEPIPYEFMA; Positive Samples: HT-29, Raji, BxPC-3, DU145, Mouse liver.
Specifications
Specifications
| Antigen | RPL21 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Rpl21 |
| Gene Accession No. | O09167, P20280, P46778 |
| Gene Alias | 60S ribosomal protein L21; 8430440E03Rik; HYPT12; L21; Large ribosomal subunit protein eL21; ribosomal protein L21; ribosomal protein L21 pseudogene 16; Rpl21; RPL21P16; similar to ribosomal protein L21 |
| Gene Symbols | Rpl21 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.