Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL22L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584777 Supplier Invitrogen™ Supplier No. PA584777
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFSKRYLKYL TKKYLKKNNL RDWLRVVAS Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Ribosomal Protein L22 is also known as heparin-binding protein HBp15, because it binds heparin in the submandibular gland and brain. This small protein is also associated with two small nuclear RNAs called EBERs (Epstein- Barr encoded RNAs). These RNAs are synthesized in large amounts by human B lymphocytes infected with Epstein-Barr virus (EBV). Ribosomal protein L22, like L4, contains a globular domain that sits on the surface of the large ribosomal subunit and an extended loop that penetrates its core. These extensions contact multiple domains of 23S rRNA, indicating a potential, but not essential, role in rRNA folding during ribosomal assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL22L1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL22L1
Gene Accession No. Q6P5R6
Gene Alias 60S ribosomal protein L22-like 1; EAP; Large ribosomal subunit protein eL22-like 1; RGD1309517; ribosomal protein L22 like 1; ribosomal protein L22-like 1; RPL22; RPL22L1
Gene Symbols RPL22L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL22L1. Recombinant protein control fragment (Product #RP-108539).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.