Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL22L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575700
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ribosomal Protein L22 is also known as heparin-binding protein HBp15, because it binds heparin in the submandibular gland and brain. This small protein is also associated with two small nuclear RNAs called EBERs (Epstein- Barr encoded RNAs). These RNAs are synthesized in large amounts by human B lymphocytes infected with Epstein-Barr virus (EBV). Ribosomal protein L22, like L4, contains a globular domain that sits on the surface of the large ribosomal subunit and an extended loop that penetrates its core. These extensions contact multiple domains of 23S rRNA, indicating a potential, but not essential, role in rRNA folding during ribosomal assembly.
Specifications
RPL22L1 | |
Polyclonal | |
Unconjugated | |
RPL22L1 | |
60S ribosomal protein L22-like 1; EAP; Large ribosomal subunit protein eL22-like 1; RGD1309517; ribosomal protein L22 like 1; ribosomal protein L22-like 1; RPL22; RPL22L1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200916, 361923 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P5R6 | |
RPL22L1 | |
Synthetic peptide corresponding to residues in Human RPL22. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction