Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551916 Supplier Invitrogen™ Supplier No. PA551916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISLGLPVGAV INCADNTGAK NLYIISVKGI KGRLNRLPAA GVGDMVMATV KKGKPELRKK VHPAVVIRQR KSYRRKDGV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL23
Gene Accession No. P62829
Gene Alias 2810009A01Rik; 60S ribosomal protein L17; 60S ribosomal protein L23; L23; Large ribosomal subunit protein uL14; MGC111167; MGC117346; MGC72008; ribosomal protein L17; ribosomal protein L23; rpL17; Rpl23
Gene Symbols RPL23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL23. Recombinant protein control fragment (Product #RP-102216).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.