Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL23A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562473 Supplier Invitrogen™ Supplier No. PA562473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNTLVFTVDV KANKHQIRQA VKKLYD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL23A is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L23P family of ribosomal proteins. It is located in the cytoplasm. The protein may be one of the target molecules involved in mediating growth inhibition by interferon. In yeast, the corresponding protein binds to a specific site on the 26S rRNA. This gene is co-transcribed with the U42A, U42B, U101A, and U101B small nucleolar RNA genes, which are located in its third, first, second, and fourth introns, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL23A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL23A
Gene Accession No. P62750
Gene Alias 60S ribosomal protein L23a; BC029892; L23A; Large ribosomal subunit protein uL23; MDA20; melanoma differentiation-associated gene 20; ribosomal protein L23a; RPL23A
Gene Symbols RPL23A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL23A. Recombinant protein control fragment (Product #RP-98945).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.