Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589707 Supplier Invitrogen™ Supplier No. PA589707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAFLSKRNPR QINWTVLYRR KHKKGQSEEI QKKRTRRAVK FQRAITGASL ADIMAKRNQK PEVRKAQREQ AIRAAKEAKK AKQASKKTAM AAAKAPTKAA PKQKIVKPVK VSAPRVGGKR; Positive Samples: HT-29, HepG2, HeLa.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L24E family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as ribosomal protein L30 because the encoded protein shares amino acid identity with the L30 ribosomal proteins from S. cerevisiae; however, its official name is ribosomal protein L24. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL24
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Rpl24
Gene Accession No. P83731, P83732, Q8BP67
Gene Alias 0610008L05Rik; 60S ribosomal protein L24; 60S ribosomal protein L30; belly spot and tail; Bst; epididymis secretory protein Li 310; HEL-S-310; L24; L30; Large ribosomal subunit protein eL24; ribosomal protein L24; ribosomal protein L30; Rpl24
Gene Symbols Rpl24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 38-157 of human RPL24 (NP_0009771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6152, 64307, 68193
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.