Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103542
Description
Antibody detects endogenous levels of total RPL24.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L24E family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as ribosomal protein L30 because the encoded protein shares amino acid identity with the L30 ribosomal proteins from S. cerevisiae; however, its official name is ribosomal protein L24. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPL24 | |
Polyclonal | |
Unconjugated | |
Rpl24 | |
0610008L05Rik; 60S ribosomal protein L24; 60S ribosomal protein L30; belly spot and tail; Bst; epididymis secretory protein Li 310; HEL-S-310; L24; L30; Large ribosomal subunit protein eL24; ribosomal protein L24; ribosomal protein L30; Rpl24 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6152, 64307, 68193 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P83731, P83732, Q8BP67 | |
Rpl24 | |
A synthesized peptide derived from human RPL24(Accession P83731), corresponding to amino acid residues R73-K123. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction