Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL26L1 Antibody (SR1266), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32251950UL
Description
RPL26L1 Monoclonal antibody specifically detects RPL26L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
RPL26L1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1266 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
FLJ46904, ribosomal protein L26 homolog, ribosomal protein L26 pseudogene 1, ribosomal protein L26-like 1, RPL26P1,60S ribosomal protein L26-like 1 | |
A synthesized peptide derived from human RPL26L1 (Uniprot #: Q9UNX3) | |
50 μg | |
Cell Biology | |
51121 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction