Learn More
Invitrogen™ RPL26L1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total RPL26L1 protein.
Specifications
Specifications
| Antigen | RPL26L1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPL26L1 |
| Gene Accession No. | Q9UNX3 |
| Gene Alias | 60S ribosomal protein L26-like 1; Large ribosomal subunit protein uL24-like 1; ribosomal protein L26 homolog; ribosomal protein L26 like 1; ribosomal protein L26 pseudogene 1; RPL26L1; RPL26P1 |
| Gene Symbols | Rpl26-ps3, RPL26L1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.