Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL26L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597191 Supplier Invitrogen™ Supplier No. PA597191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKFNPFVTSD RSKNRKRHFN APSHVRRKIM SSPLSKELRQ KYNVRSMPIR KDDEVQVVRG HYKGQQIGKV VQVYRKKYVI YIERVQREKA NGTTVHVGIH PSKVVITRLK LDKDRKKILE RKAKSRQVGK EKGKYKEELI EKMQE; Positive Samples: Jurkat, 293T, HepG2, HeLa.

This gene encodes a protein that shares high sequence similarity with ribosomal protein L26. It is not currently known whether the encoded protein is a functional ribosomal protein or whether it has evolved a function that is independent of the ribosome. Transcript variants utilizing alternative polyA signals exist. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL26L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPL26L1
Gene Accession No. Q9UNX3
Gene Alias 60S ribosomal protein L26-like 1; Large ribosomal subunit protein uL24-like 1; ribosomal protein L26 homolog; ribosomal protein L26 like 1; ribosomal protein L26 pseudogene 1; RPL26L1; RPL26P1
Gene Symbols RPL26L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human RPL26L1 (NP_0571771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51121
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.