Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL27A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563896 Supplier Invitrogen™ Supplier No. PA563896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INFDKYHPGY FGKVGMKHYH LKRNQSFCPT VNLDKLWTLV SEQTRVNAAK NKTGAAPIID VVRSGYYKVL G Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Rpl27a gene ontology annotations related to this gene include translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL27A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL27A
Gene Accession No. P46776
Gene Alias 60S ribosomal protein L27a; 60S ribosomal protein L27-A; 60S ribosomal protein L27a-like protein; fa93g12; FLJ43464; hypothetical protein LOC336726; L27'; L27A; L29; Large ribosomal subunit protein eL27-A; Large ribosomal subunit protein uL15; MGC10850; MGC17878; MGC87238; ORF; ORF protein; ribosomal 60S subunit protein L27A; ribosomal protein L27A; ribosomal protein L27a-like; ribosomal protein L29 homolog; RPL27; RPL27A; wu:fa93g12; YHR010W; zgc:77235
Gene Symbols RPL27A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL27A. Recombinant protein control fragment (Product #RP-103382).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.