Learn More
Invitrogen™ RPL29 Polyclonal Antibody

Description
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (84%), Rat (85%), Pig (93%), Rhesus Monkey (97%), Chimpanzee (96%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RPL29 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RPL29 |
| Gene Accession No. | P47914 |
| Gene Alias | 60S ribosomal protein L29; cell surface heparin-binding protein HIP; heparin/heparan sulfate-binding protein; heparin/heparan sulfate-interacting protein; HIP; HP/HS-interacting protein; HUMRPL29; L29; Large ribosomal subunit protein eL29; P23; ribosomal protein L29; ribosomal protein YL43 homologue; Rpl29; RPL29_3_370; RPL29P10; Rpl43 |
| Gene Symbols | RPL29 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.