Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116756 Supplier Invitrogen™ Supplier No. PA5116756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, HepG2, Mouse spleen, Mouse brain, Mouse liver, Rat spleen Immunogen sequence: KKGGEKKKGR SAINEVVTRE YTINIHKRIH GVGFKKRAPR ALKEIRKFAM KEMGTPDVRI DTRLNKAVWA KGIRNVPYRI RVRLSRKRNE DEDSPNKLYT L.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL31 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L31E family of ribosomal proteins. It is located in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL31
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPL31
Gene Accession No. P62899, P62900, P62902
Gene Alias 60S ribosomal protein L31; L31; Large ribosomal subunit protein eL31; ribosomal protein L31; RPL31
Gene Symbols RPL31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 5-105 of human RPL31 (NP_0009841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114641, 6160, 64298
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.