Learn More
Invitrogen™ RPL32 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (91%), Xenopus laevis (92%), Dog (100%), Pig (100%), Rabbit (94%), Chicken (94%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RPL32 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Rpl32 |
| Gene Accession No. | P62910 |
| Gene Alias | 60S ribosomal protein L32; AU020185; hypothetical protein LOC615556; hypothetical protein LOC677748; L32; Large ribosomal subunit protein eL32; PP9932; ribosomal protein L32; RPL32; rpL32-3A; snoRNA MBI-141; zgc:136952 |
| Gene Symbols | Rpl32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.