Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120387 Supplier Invitrogen™ Supplier No. PA5120387
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse liver. Immunogen sequence: MVQRLTYRRR LSYNTASNKT RLSRTPGNRI VYLYTKKVGK APKSACGVCP GRLRGVRAVR PKVLMRLSKT KKHVSRAYGG SMCAKCVRDR IKRAFLIEEQ KIVVKVLKAQ AQSQKAK.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L34E family of ribosomal proteins. It is located in the cytoplasm. This gene originally was thought to be located at 17q21, but it has been mapped to 4q. Transcript variants derived from alternative splicing, alternative transcription initiation sites, and/or alternative polyadenylation exist; these variants encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL34
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPL34
Gene Accession No. P49207, Q9D1R9
Gene Alias 1100001I22Rik; 60S ribosomal protein L34; L34; Large ribosomal subunit protein eL34; leukemia-associated protein; ribosomal protein L34; ribosomal protein L34-like 2; RL34; RPL34; Rpl34l2
Gene Symbols RPL34
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human RPL34 (NP_000986.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6164, 68436
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.