Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL35A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110319
Description
Immunogen sequence: IFAGYKRGLR NQREHTALLK IEGVYARDET EFYLGKRCAY VYKAKNNTVT PGGKPNKTRV IWGKVTRAHG NSGMVRAKFR S.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L35AE family of ribosomal proteins. It is located in the cytoplasm. The rat protein has been shown to bind to both initiator and elongator tRNAs, and thus, it is located at the P site, or P and A sites, of the ribosome.
Specifications
| RPL35A | |
| Polyclonal | |
| Unconjugated | |
| RPL35A | |
| 2810431L15Rik; 60S ribosomal protein L35a; cell growth-inhibiting gene 33 protein; DBA5; GIG33; L35A; Large ribosomal subunit protein eL33; ribosomal protein L35A; Rpl35; RPL35A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57808, 57809 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.21 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O55142, P04646 | |
| RPL35A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-90 of human RPL35A (NP_000987.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction