Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL36AL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110320
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110320 Supplier Invitrogen™ Supplier No. PA5110320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVNVPKTRRT FCKKCGKHQP HKVTQYKKGK DSLYAQGRRR YDRKQSGYGG QTKPIFRKKA KTTKKIVLRL ECVEPNCRSK RMLAIKRCKH FELGGDKKRK GQVIQF.

Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with yeast ribosomal protein L44, belongs to the L44E (L36AE) family of ribosomal proteins. This gene and the human gene officially named ribosomal protein L36a (RPL36A) encode nearly identical proteins; however, they are distinct genes. Although the name of this gene has been referred to as ribosomal protein L36a (RPL36A), its official name is ribosomal protein L36a-like (RPL36AL). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL36AL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPL36AL
Gene Accession No. Q969Q0
Gene Alias 2410038A03Rik; 60S ribosomal protein L36a; 60S ribosomal protein L36a-like; 60S ribosomal protein L44; Large ribosomal subunit protein eL42-like; large subunit ribosomal protein L36a; MGC111574; ribosomal protein HL44; ribosomal protein L36a; ribosomal protein L36a like; ribosomal protein L36a-like; Rpl36a; RPL36A protein; RPL36AL; Rpl44
Gene Symbols RPL36AL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-106 of human RPL36AL (NP_000992.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6166, 66483, 81769
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.