Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL36AL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112652
Description
RPL36AL Polyclonal Antibody for Western Blot, ELISA
Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with yeast ribosomal protein L44, belongs to the L44E (L36AE) family of ribosomal proteins. This gene and the human gene officially named ribosomal protein L36a (RPL36A) encode nearly identical proteins; however, they are distinct genes. Although the name of this gene has been referred to as ribosomal protein L36a (RPL36A), its official name is ribosomal protein L36a-like (RPL36AL). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL36AL | |
| Polyclonal | |
| Unconjugated | |
| RPL36AL | |
| 2410038A03Rik; 60S ribosomal protein L36a; 60S ribosomal protein L36a-like; 60S ribosomal protein L44; Large ribosomal subunit protein eL42-like; large subunit ribosomal protein L36a; MGC111574; ribosomal protein HL44; ribosomal protein L36a; ribosomal protein L36a like; ribosomal protein L36a-like; Rpl36a; RPL36A protein; RPL36AL; Rpl44 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 6166 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q969Q0 | |
| RPL36AL | |
| Recombinant Human 60S ribosomal protein L36a-like protein (1-106AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction