Learn More
Invitrogen™ RPL37 Polyclonal Antibody

Description
Positive Samples: Jurkat, 293T, MCF-7, HeLa Immunogen sequence: MTKGTSSFGK RRNKTHTLCR RCGSKAYHLQ KSTCGKCGYP AKRKRKYNWS AKAKRRNTTG TGRMRHLKIV YRRFRHGFRE GTTPKPKRAA VAASSSS.
Specifications
Specifications
| Antigen | RPL37 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.91 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RPL37 |
| Gene Accession No. | P61927, P61928, Q9D823 |
| Gene Alias | 3110005M08Rik; 60S ribosomal protein L37; 60S ribosomal protein L37a; DKFZp686G1699; G1.16; L37; Large ribosomal subunit protein eL37; MGC99572; ribosomal protein L37; RPL37 |
| Gene Symbols | RPL37 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.