Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575865
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L37E family of ribosomal proteins. It is located in the cytoplasm. The protein contains a C2C2-type zinc finger-like motif.
Specifications
RPL37 | |
Polyclonal | |
Unconjugated | |
RPL37 | |
3110005M08Rik; 60S ribosomal protein L37; 60S ribosomal protein L37a; DKFZp686G1699; G1.16; L37; Large ribosomal subunit protein eL37; MGC99572; ribosomal protein L37; RPL37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6167, 67281, 81770 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.3 | |
P61927, P61928, Q9D823 | |
RPL37 | |
Synthetic peptide corresponding to residues in Human RPL37. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction