Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588313 Supplier Invitrogen™ Supplier No. PA588313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPRKIEEIKD FLLTARRKDA KSVKIKKNKD NVKFKVRCSR YLYTLVITDK EKAEKLKQSL PPGLAVKELK; Positive Samples: A-375, HeLa, MCF-7.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L38E family of ribosomal proteins. It is located in the cytoplasm. Alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome, including one located in the promoter region of the type 1 angiotensin II receptor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL38
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPL38
Gene Accession No. P63173
Gene Alias 0610025G13Rik; 60S ribosomal protein L38; L38; Large ribosomal subunit protein eL38; Rbt; ribosomal protein L38; Rpl38; Ts; Tss
Gene Symbols RPL38
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human RPL38 (NP_0009901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6169
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.