Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL39 Polyclonal Antibody
RPL39 Polyclonal Antibody
Supplier: Thermo Scientific PA5101390
Description
RPL39 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
RPL39 | |
Polyclonal | |
Unconjugated | |
Rpl39 | |
2810465O16Rik; 60S ribosomal protein L39; L39; Large ribosomal subunit protein eL39; ribosomal protein L39; Rpl39; RPL39_23_1806; RPL39P42 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25347, 6170, 67248 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62891, P62892, P62893 | |
Rpl39 | |
A synthesized peptide derived from human RPL39, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction