Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL39L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561427 Supplier Thermo Scientific Supplier No. PA561427
Add to Cart
Add to Cart

RPL39 Polyclonal Antibody

RPL39 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL39L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL39L
Gene Accession No. Q96EH5
Gene Alias 60S ribosomal protein L39-2; 60S ribosomal protein L39-like; L39-2; Large ribosomal subunit protein eL39-like; ribosomal protein L39 like; ribosomal protein L39-like 1; ribosomal protein L39-like protein; RPL39L; RPL39L1
Gene Symbols RPL39L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL39. Recombinant protein control fragment (Product #RP-100800).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.