Learn More
Invitrogen™ RPL3L Polyclonal Antibody

Description
The antibody detects endogenous levels of total RPL3L protein.
Specifications
Specifications
| Antigen | RPL3L |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPL3L |
| Gene Accession No. | Q92901 |
| Gene Alias | 1110057H16Rik; 60S ribosomal protein L3-like; Large ribosomal subunit protein uL3-like; ribosomal protein L3 like; ribosomal protein L3 pseudogene; ribosomal protein L3-like; RPL3L |
| Gene Symbols | RPL3L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.