Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561891 Supplier Invitrogen™ Supplier No. PA561891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHGRFQTAQE KRAFMGPQKK HLEKETPETS GDL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

A full-length cDNA encoding a novel ribosomal protein L3 gene was isolated and sequenced. The deduced protein sequence is 407 amino acids long and shows 77% identity to other known mammalian ribosomal protein L3 genes, which are themselves highly conserved. Southern blot analysis of human genomic DNA suggests that this novel gene is single copy. While the previously identified human ribosomal protein L3 gene has ubiquitous expression in all tissues surveyed, the novel gene described herein is strongly expressed in skeletal muscle and heart tissue, with low levels of expression in the pancreas. This novel gene, RPL3L, is located in a gene-rich region near the PKD1 and TSC2 genes on chromosome 16p13.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL3L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL3L
Gene Accession No. Q92901
Gene Alias 1110057H16Rik; 60S ribosomal protein L3-like; Large ribosomal subunit protein uL3-like; ribosomal protein L3 like; ribosomal protein L3 pseudogene; ribosomal protein L3-like; RPL3L
Gene Symbols RPL3L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL3L. Recombinant protein control fragment (Product #RP-100667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.