Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL3L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575763
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
A full-length cDNA encoding a novel ribosomal protein L3 gene was isolated and sequenced. The deduced protein sequence is 407 amino acids long and shows 77% identity to other known mammalian ribosomal protein L3 genes, which are themselves highly conserved. Southern blot analysis of human genomic DNA suggests that this novel gene is single copy. While the previously identified human ribosomal protein L3 gene has ubiquitous expression in all tissues surveyed, the novel gene described herein is strongly expressed in skeletal muscle and heart tissue, with low levels of expression in the pancreas. This novel gene, RPL3L, is located in a gene-rich region near the PKD1 and TSC2 genes on chromosome 16p13.3.
Specifications
RPL3L | |
Polyclonal | |
Unconjugated | |
RPL3L | |
1110057H16Rik; 60S ribosomal protein L3-like; Large ribosomal subunit protein uL3-like; ribosomal protein L3 like; ribosomal protein L3 pseudogene; ribosomal protein L3-like; RPL3L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287122, 6123, 66211 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92901 | |
RPL3L | |
Synthetic peptide corresponding to residues in Human RPL3L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction