Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576894
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L4E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL4 | |
| Polyclonal | |
| Unconjugated | |
| RPL4 | |
| 2010004J23Rik; 60S ribosomal protein L1; 60S ribosomal protein L4; fa97a02; fl04b10; L1; L4; Large ribosomal subunit protein uL4; ribosomal protein L4; RPL1; RPL4; wu:fa97a02; wu:fa97c01; wu:fl04b10; zgc:56659; zgc:85693 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6124, 64302, 67891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P36578, P50878, Q9D8E6 | |
| RPL4 | |
| Recombinant full length Human RPL4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction