Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598127
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598127 Supplier Invitrogen™ Supplier No. PA598127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L4E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RPL4
Gene Accession No. P36578
Gene Alias 2010004J23Rik; 60S ribosomal protein L1; 60S ribosomal protein L4; fa97a02; fl04b10; L1; L4; Large ribosomal subunit protein uL4; ribosomal protein L4; RPL1; RPL4; wu:fa97a02; wu:fa97c01; wu:fl04b10; zgc:56659; zgc:85693
Gene Symbols RPL4
Host Species Rabbit
Immunogen Recombinant Human 60S ribosomal protein L4 protein (12-91AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6124
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.