Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103549 Supplier Invitrogen™ Supplier No. PA5103549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPL41.

Interacts with the beta subunit of protein kinase CKII and stimulates phosphorylation of DNA topoisomerase II alpha by CKII.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL41
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPL41
Gene Accession No. P62945, P62947, P62948
Gene Alias 1810055P16Rik; 2210411K19Rik; 60S ribosomal protein L41; HG12; HG12 protein; homologue of yeast ribosomal protein YL41; L41; Large ribosomal subunit protein eL41; ribosomal protein L41; Rpl41
Gene Symbols RPL41
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPL41(Accession P62945), corresponding to amino acid residues M1-K25.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124440, 6171, 67945
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.