Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562750 Supplier Invitrogen™ Supplier No. PA562750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKWRKKRMRR LKRKRRKMRQ RSK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Interacts with the beta subunit of protein kinase CKII and stimulates phosphorylation of DNA topoisomerase II alpha by CKII.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL41
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL41
Gene Accession No. P62945
Gene Alias 1810055P16Rik; 2210411K19Rik; 60S ribosomal protein L41; HG12; HG12 protein; homologue of yeast ribosomal protein YL41; L41; Large ribosomal subunit protein eL41; ribosomal protein L41; Rpl41
Gene Symbols RPL41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL41. Recombinant protein control fragment (Product #RP-100652).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.