Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560481 Supplier Invitrogen™ Supplier No. PA560481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLNRFGMDK IYEGQVEVTG DEYNVESIDG QPGAFTCYLD AGLARTTTGN KVFGALKGAV DGGLSIPHST KRFPGYDSES KEFNAEV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

RPL5 encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L18P family of ribosomal proteins. It is located in the cytoplasm. The protein binds 5S rRNA to form a stable complex called the 5S ribonucleoprotein particle (RNP), which is necessary for the transport of nonribosome-associated cytoplasmic 5S rRNA to the nucleolus for assembly into ribosomes. The protein interacts specifically with the beta subunit of casein kinase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL5
Gene Accession No. P09895, P46777, P47962
Gene Alias 60S ribosomal protein L5; DBA6; L5; Large ribosomal subunit protein uL18; MSTP030; PPP1R135; protein phosphatase 1, regulatory subunit 135; ribosomal protein L5; RPL5; U21RNA
Gene Symbols RPL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL5. Recombinant protein control fragment (Product #RP-100628).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100503670, 6125, 81763
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.