Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111511 Supplier Invitrogen™ Supplier No. PA5111511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRVPLRRTHQ KFVIATSTKI DISNVKIPKH LTDAYFKKKK LRKPRHQEGE IF Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6E family of ribosomal proteins. It is located in the cytoplasm. The protein can bind specifically to domain C of the tax-responsive enhancer element of human T-cell leukemia virus type 1, and it has been suggested that the protein may participate in tax-mediated transactivation of transcription. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL6
Gene Accession No. Q02878
Gene Alias 60S ribosomal protein L6; DNA-binding protein TAXREB107; L6; Large ribosomal subunit protein eL6; Neoplasm-related protein C140; ribosomal protein L6; Rpl6; SHUJUN-2; Taxreb107; TAX-responsive enhancer element-binding protein 107; TXREB1
Gene Symbols RPL6
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human RPL6. Recombinant protein control fragment (Product #RP-104075).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.