Learn More
RPL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 153871AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0193
Only null left
Add to Cart
Description
Description
This gene encodes a protein component of the 60S ribosomal subunit. This protein can bind specifically to domain C of the tax-responsive enhancer element of human T-cell leukemia virus type 1, and may participate in tax-mediated transactivation of transcription. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
RPL6
Polyclonal
Unconjugated
RPL6
60S ribosomal protein L6, Neoplasm related protein C140, ribosomal protein L6, RPL6, SHUJUN 2, TAXREB107, TXREB1
Rabbit
Antigen Affinity Chromatography
RUO
117042, 19988, 6128
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21533, P47911, Q02878
RPL6
RPL6 Fusion Protein Ag6840
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.