Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL7 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002268AF594
Description
RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot.Specifications
RPL7 | |
Polyclonal | |
Western Blot | |
humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7 | |
Rabbit | |
Affinity Purified | |
Immune System Diseases, Immunology | |
6129.0 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
RPL7 | |
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction