Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL7A Recombinant Rabbit Monoclonal Antibody (4I8Y6)
GREENER_CHOICE

Catalog No. PIMA537881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537881 Supplier Invitrogen™ Supplier No. MA537881
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: 293T, PC-12, HepG2, NIH/3T3, Raji, Mouse lung, Mouse liver, Rat lung, Rat liver Immunogen sequence: MPKGKKAKGK KVAPAPAVVK KQEAKKVVNP LFEKRPKNFG IGQDIQPKRD LTRFVKWPRY IRLQRQRAIL YKRLKVPPAI NQFTQALDRQ TATQLLKLAH KYRPETKQEK KQRLLARAEK KAAGKGDVPT KRPPVLRAGV NTVTTLVENK KAQLVVIAHD VDPIE.

Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L7AE family of ribosomal proteins. It can interact with a subclass of nuclear hormone receptors, including thyroid hormone receptor, and inhibit their ability to transactivate by preventing their binding to their DNA response elements. This gene is included in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. It is co-transcribed with the U24, U36a, U36b, and U36c small nucleolar RNA genes, which are located in its second, fifth, fourth, and sixth introns, respectively. This gene rearranges with the trk proto-oncogene to form the chimeric oncogene trk-2h, which encodes an oncoprotein consisting of the N terminus of ribosomal protein L7a fused to the receptor tyrosine kinase domain of trk. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL7A
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4I8Y6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rpl7a
Gene Accession No. P12970, P62424, P62425
Gene Alias 60S ribosomal protein L7a; L7A; Large ribosomal subunit protein eL8; PLA-X polypeptide; ribosomal protein L7a; RPL7A; Surf3; SURF-3; surfeit 3; surfeit locus protein 3; thyroid hormone receptor uncoupling protein; TRUP
Gene Symbols Rpl7a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human RPL7A (P62424).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27176, 296596, 6130
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.