Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541712
Description
Peptide sequence: GVACSVSQAQ KDELILEGND IELVSNSAAL IQQATTVKNK DIRKFLDGIY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
RPL9 | |
Polyclonal | |
Unconjugated | |
RPL9 | |
60S ribosomal protein L9; 60S ribosomal protein L9-like protein; ab02c03; fa93a01; L9; Large ribosomal subunit protein uL6; mg:ab02c03; NPC-A-16; OK/SW-cl.103; ribosomal protein L9; ribosomal protein L9-like; ribosomal protein L9-like protein; RPL9; rpl9 protein; RPL9P7; RPL9P8; RPL9P9; similar to ribosomal protein L9; wu:fa93a01; zgc:103730 | |
Rabbit | |
Protein A | |
RUO | |
6133 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P32969 | |
RPL9 | |
Synthetic peptide directed towards the C-terminal of human RPL9 (aa 131-180). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction