Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598129 Supplier Invitrogen™ Supplier No. PA598129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL9 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPL9
Gene Accession No. P32969
Gene Alias 60S ribosomal protein L9; 60S ribosomal protein L9-like protein; ab02c03; fa93a01; L9; Large ribosomal subunit protein uL6; mg:ab02c03; NPC-A-16; OK/SW-cl.103; ribosomal protein L9; ribosomal protein L9-like; ribosomal protein L9-like protein; RPL9; rpl9 protein; RPL9P7; RPL9P8; RPL9P9; similar to ribosomal protein L9; wu:fa93a01; zgc:103730
Gene Symbols RPL9
Host Species Rabbit
Immunogen Recombinant Human 60S ribosomal protein L9 protein (1-192AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6133
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.