Learn More
Invitrogen™ RPLP0 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589335
Description
Immunogen sequence: MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLVIQ QVFDNGSIYN PEVLDITEET LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETDYTFP LAEKVKAFLA DPSAFVAAAP VAAATTAAPA AAAAPAKVEA KEESEESDED MGFGLFD; Positive Samples: SW620, OVCAR3, Mouse liver, Mouse thymus; Cellular Location: Cytoplasm, Nucleus.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which is the functional equivalent of the E. coli L10 ribosomal protein, belongs to the L10P family of ribosomal proteins. It is a neutral phosphoprotein with a C-terminal end that is nearly identical to the C-terminal ends of the acidic ribosomal phosphoproteins P1 and P2. The P0 protein can interact with P1 and P2 to form a pentameric complex consisting of P1 and P2 dimers, and a P0 monomer. The protein is located in the cytoplasm. Transcript variants derived from alternative splicing exist; they encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPLP0 | |
Polyclonal | |
Unconjugated | |
RPLP0 | |
36B4; 60S acidic ribosomal protein P0; 60S acidic ribosomal protein P0-like protein; 60S ribosomal protein L10E; acidic ribosomal phophoprotein; acidic ribosomal phophoprotein P0; acidic ribosomal phosphoprotein P0; acidic ribosomal phosphoprotein PO; arbp; arp; fa56b12; fb04a12; L10E; Large ribosomal subunit protein uL10; LP0; MGC107165; MGC107166; MGC111226; MGC88175; neutral ribosomal phosphoprotein P0; OTTMUSP00000015585; P0; PRLP0; ribosomal protein lateral stalk subunit P0; ribosomal protein P0; ribosomal protein, large, P0; RPLP0; rplp0 protein; rplRP; RPP0; similar to acidic ribosomal phosphoprotein PO; wu:fa56b12; wu:fb15a08; wu:fk48c12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11837, 6175 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05388, P14869 | |
RPLP0 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human RPLP0 (NP_0009931). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.