Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPLP0 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984263
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4263 Supplier Bioss Supplier No. BS1886R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which is the functional equivalent of the E. coli L10 ribosomal protein, belongs to the L10P family of ribosomal proteins. It is a neutral phosphoprotein with a C-terminal end that is nearly identical to the C-terminal ends of the acidic ribosomal phosphoproteins P1 and P2. The P0 protein can interact with P1 and P2 to form a pentameric complex consisting of P1 and P2 dimers, and a P0 monomer. The protein is located in the cytoplasm. Transcript variants derived from alternative splicing exist; they encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.

Specifications

Antigen RPLP0
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RPLP0
Gene Accession No. P05388
Gene Alias 36B4; 60S acidic ribosomal protein P0; 60S acidic ribosomal protein P0-like protein; 60S ribosomal protein L10E; acidic ribosomal phophoprotein; acidic ribosomal phophoprotein P0; acidic ribosomal phosphoprotein P0; Arbp; arp; BOS_16487; fa56b12; fb04a12; L10E; Large ribosomal subunit protein uL10; LP0; MGC107165; MGC107166; MGC111226; MGC88175; neutral ribosomal phosphoprotein P0; OTTMUSP00000015585; P0; PRLP0; ribosomal protein lateral stalk subunit P0; ribosomal protein P0; ribosomal protein, large, P0; RPLP0; rplp0 protein; rplRP; RPP0; wu:fa56b12; wu:fb15a08; wu:fk48c12
Gene Symbols RPLP0
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RPLP0.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6175
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.