Learn More
Invitrogen™ RPLP1 Monoclonal Antibody (1G10)

Description
Peptide Sequence: MASVSELACI YSALILHDDE VTVTEDKINA LIKAAGVNVE PFWPGLFAKA LANVNIGSLI CNVGAGGPAP AAGAAPAGGP APSTAAAPAE EKKVEAKKEE SEESDDDMGF GLFD.
Specifications
Specifications
| Antigen | RPLP1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1G10 |
| Concentration | 0.47 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RPLP1 |
| Gene Accession No. | P05386 |
| Gene Alias | 2410042H16Rik; 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; Arpp1; C430017H15Rik; L RPP1; Large ribosomal subunit protein P1; LP1; P1; ribosomal protein lateral stalk subunit P1; ribosomal protein, large, P1; RPLP1; RPP1; RRP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.